Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFA2A
Entrez Gene ID
286882
Gene Name
BPI fold containing family A, member 2A
Gene Aliases
BSP30A, SPLUNC2A
Chromosome Location
Chr.13: 62503682 - 62514221 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174803.3 NP_777228.2 6 - 7 723 84
GenBank mRNA U79413.2 - 6 - 7 723 84

View Full Product Details