Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEMT
Entrez Gene ID
360197
Gene Name
phosphatidylethanolamine N-methyltransferase
Gene Aliases
PLMT
Chromosome Location
Chr.19: 34779429 - 34816445 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182989.3 NP_892034.1 8 - 9 997 76
GenBank mRNA BC105466.1 - 8 - 9 998 76

View Full Product Details