Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTF1
Entrez Gene ID
509960
Gene Name
metal regulatory transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.3: 108062883 - 108103486 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035080.2 NP_001030252.2 3 - 4 739 75
GenBank mRNA AY309439.1 - 2 - 3 650 75
BC123774.1 - 3 - 4 739 75
BT020946.1 - 3 - 4 769 75

View Full Product Details