Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM41A
Entrez Gene ID
505368
Gene Name
transmembrane protein 41A
Gene Aliases
-
Chromosome Location
Chr.1: 81696439 - 81705118 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075200.1 NP_001068668.1 1 - 2 174 95
GenBank mRNA BC123868.1 - 1 - 2 174 95

View Full Product Details