The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001075206.1 | NP_001068674.1 | 10 - 11 | 1718 | 81 |
GenBank mRNA | BT026224.1 | - | 10 - 11 | 1718 | 81 |