Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VWA7
Entrez Gene ID
505555
Gene Name
von Willebrand factor A domain containing 7
Gene Aliases
C23H6orf27, C6ORF27, G7C
Chromosome Location
Chr.23: 27546263 - 27555824 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075206.1 NP_001068674.1 8 - 9 1411 62
GenBank mRNA BT026224.1 - 8 - 9 1411 62

View Full Product Details