Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOGA1
Entrez Gene ID
506001
Gene Name
4-hydroxy-2-oxoglutarate aldolase 1
Gene Aliases
C10orf65, DHDPSL, NPL2
Chromosome Location
Chr.26: 18771696 - 18793851 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075237.1 NP_001068705.1 2 - 3 716 81
GenBank mRNA BC119998.1 - 2 - 3 370 81
BT026547.1 - 2 - 3 716 81

View Full Product Details