The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001075309.1 | NP_001068777.1 | 2 - 3 | 594 | 65 |
GenBank mRNA | BC119902.1 | - | 2 - 3 | 594 | 65 |