Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD16
Entrez Gene ID
507779
Gene Name
ankyrin repeat domain 16
Gene Aliases
-
Chromosome Location
Chr.13: 42879315 - 42914516 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075331.1 NP_001068799.1 5 - 6 867 67
GenBank mRNA BC120321.1 - 5 - 6 867 67

View Full Product Details