Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPO
Entrez Gene ID
508767
Gene Name
carboxypeptidase O
Gene Aliases
-
Chromosome Location
Chr.2: 95234725 - 95266753 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075372.2 NP_001068840.1 4 - 5 422 82
GenBank mRNA BC122774.1 - 4 - 5 423 82

View Full Product Details