Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDDC2
Entrez Gene ID
509282
Gene Name
HD domain containing 2
Gene Aliases
-
Chromosome Location
Chr.9: 25960234 - 25979949 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075401.1 NP_001068869.1 2 - 3 239 79
GenBank mRNA BC120449.1 - 2 - 3 239 79

View Full Product Details