The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001075405.1 | NP_001068873.1 | 1 - 2 | 295 | 88 |
GenBank mRNA | BC116005.1 | - | 1 - 2 | 313 | 88 |