Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRGC4
Entrez Gene ID
509614
Gene Name
T cell receptor gamma C4
Gene Aliases
TARP
Chromosome Location
Chr.4: 82720193 - 82746245 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC120033.1 - 3 - 4 450 130

View Full Product Details