Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DUS2
Entrez Gene ID
509795
Gene Name
dihydrouridine synthase 2
Gene Aliases
DUS2L
Chromosome Location
Chr.18: 35468636 - 35511999 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075419.1 NP_001068887.1 10 - 11 683 65
GenBank mRNA BC120342.1 - 10 - 11 683 65

View Full Product Details