Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CMTM5
Entrez Gene ID
510145
Gene Name
CKLF like MARVEL transmembrane domain containing 5
Gene Aliases
-
Chromosome Location
Chr.10: 21544233 - 21547165 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075434.1 NP_001068902.1 3 - 4 422 60
GenBank mRNA BC118492.1 - 3 - 4 422 60

View Full Product Details