Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFI35
Entrez Gene ID
510697
Gene Name
interferon induced protein 35
Gene Aliases
-
Chromosome Location
Chr.19: 43030232 - 43043997 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075462.2 NP_001068930.1 1 - 2 184 78
GenBank mRNA BC122808.1 - 1 - 2 185 78

View Full Product Details