Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFI35
Entrez Gene ID
510697
Gene Name
interferon induced protein 35
Gene Aliases
-
Chromosome Location
Chr.19: 43030232 - 43043997 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075462.2 NP_001068930.1 5 - 6 721 118
GenBank mRNA BC122808.1 - 5 - 6 722 118

View Full Product Details