Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNTTIP2
Entrez Gene ID
511258
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 49610656 - 49621986 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075506.1 NP_001068974.1 2 - 3 1724 120
GenBank mRNA BC120039.1 - 2 - 3 1724 120

View Full Product Details