Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT25
Entrez Gene ID
511540
Gene Name
keratin 25
Gene Aliases
-
Chromosome Location
Chr.19: 40945831 - 40953601 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075523.1 NP_001068991.1 2 - 3 519 70
GenBank mRNA BC119958.1 - 2 - 3 519 70

View Full Product Details