Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBL2
Entrez Gene ID
511758
Gene Name
transducin beta like 2
Gene Aliases
-
Chromosome Location
Chr.25: 33639589 - 33644862 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075535.2 NP_001069003.1 3 - 4 476 101
GenBank mRNA BC120023.1 - 3 - 4 477 101

View Full Product Details