Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STMN4
Entrez Gene ID
512732
Gene Name
stathmin 4
Gene Aliases
-
Chromosome Location
Chr.8: 74340257 - 74363975 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075580.2 NP_001069048.1 4 - 5 447 97
GenBank mRNA BC120123.1 - 4 - 5 448 97

View Full Product Details