Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOLC1
Entrez Gene ID
513278
Gene Name
nucleolar and coiled-body phosphoprotein 1
Gene Aliases
-
Chromosome Location
Chr.26: 22819792 - 22830530 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075608.2 NP_001069076.1 11 - 12 1908 109
GenBank mRNA BC122662.1 - 11 - 12 1909 109

View Full Product Details