Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOLC1
Entrez Gene ID
513278
Gene Name
nucleolar and coiled-body phosphoprotein 1
Gene Aliases
-
Chromosome Location
Chr.26: 22819792 - 22830530 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075608.2 NP_001069076.1 7 - 8 929 145
GenBank mRNA BC122662.1 - 7 - 8 930 145

View Full Product Details