Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYPL2
Entrez Gene ID
514665
Gene Name
synaptophysin like 2
Gene Aliases
-
Chromosome Location
Chr.3: 33968204 - 33981459 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075676.1 NP_001069144.1 1 - 2 219 106
GenBank mRNA BC120434.1 - 1 - 2 219 106

View Full Product Details