Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM81B
Entrez Gene ID
514910
Gene Name
family with sequence similarity 81 member B
Gene Aliases
-
Chromosome Location
Chr.7: 94687293 - 94744876 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075688.2 NP_001069156.1 7 - 8 894 148
GenBank mRNA BC122751.1 - 7 - 8 895 148

View Full Product Details