Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMD5
Entrez Gene ID
515021
Gene Name
proteasome 26S subunit, non-ATPase 5
Gene Aliases
-
Chromosome Location
Chr.8: 110196039 - 110216968 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075693.1 NP_001069161.1 4 - 5 567 145
GenBank mRNA BC120300.1 - 4 - 5 567 145

View Full Product Details