Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GHDC
Entrez Gene ID
515225
Gene Name
GH3 domain containing
Gene Aliases
D11LGP1
Chromosome Location
Chr.19: 42307202 - 42311698 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075703.1 NP_001069171.1 3 - 4 453 73
GenBank mRNA BT026535.1 - 3 - 4 453 73

View Full Product Details