Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTFP1
Entrez Gene ID
525071
Gene Name
mitochondrial fission process 1
Gene Aliases
MTP18
Chromosome Location
Chr.17: 69422829 - 69436701 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075863.2 NP_001069331.1 1 - 2 170 130
GenBank mRNA BC120145.1 - 1 - 2 171 130

View Full Product Details