Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENOPH1
Entrez Gene ID
525563
Gene Name
enolase-phosphatase 1
Gene Aliases
-
Chromosome Location
Chr.6: 97216605 - 97255730 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075875.1 NP_001069343.1 2 - 3 396 89
GenBank mRNA BC119871.1 - 2 - 3 396 89

View Full Product Details