The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001075895.2 | NP_001069363.1 | 1 - 2 | 550 | 79 |
GenBank mRNA | BC120273.1 | - | 1 - 2 | 554 | 79 |