Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR4A1
Entrez Gene ID
528390
Gene Name
nuclear receptor subfamily 4 group A member 1
Gene Aliases
-
Chromosome Location
Chr.5: 27822914 - 27839686 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075911.1 NP_001069379.1 5 - 6 1367 81
GenBank mRNA BT026269.1 - 5 - 6 1367 81

View Full Product Details