Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMCES
Entrez Gene ID
530527
Gene Name
5-hydroxymethylcytosine binding, ES cell specific
Gene Aliases
C22H3orf37
Chromosome Location
Chr.22: 59150413 - 59165191 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075934.1 NP_001069402.1 3 - 4 379 81
GenBank mRNA BC120385.1 - 3 - 4 379 81

View Full Product Details