Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HES6
Entrez Gene ID
531384
Gene Name
hes family bHLH transcription factor 6
Gene Aliases
-
Chromosome Location
Chr.3: 117549870 - 117552740 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075949.1 NP_001069417.1 1 - 2 515 83
GenBank mRNA BC118125.1 - 1 - 2 536 83

View Full Product Details