Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPP2
Entrez Gene ID
534937
Gene Name
uridine phosphorylase 2
Gene Aliases
-
Chromosome Location
Chr.2: 38224677 - 38262110 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076043.2 NP_001069511.1 6 - 7 1002 89
GenBank mRNA BC120094.1 - 6 - 7 1003 89

View Full Product Details