Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMLHE
Entrez Gene ID
535630
Gene Name
trimethyllysine hydroxylase, epsilon
Gene Aliases
-
Chromosome Location
Chr.X: 35551796 - 35602891 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076064.1 NP_001069532.1 4 - 5 747 85
GenBank mRNA BC120318.1 - 4 - 5 747 85

View Full Product Details