The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076083.1 | NP_001069551.1 | 2 - 3 | 559 | 88 |
GenBank mRNA | BC120394.1 | - | 2 - 3 | 559 | 88 |