Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PVALB
Entrez Gene ID
538603
Gene Name
parvalbumin
Gene Aliases
pvalb1
Chromosome Location
Chr.5: 75176502 - 75196138 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076114.2 NP_001069582.1 - 262 85
GenBank mRNA BC120184.1 - - 263 85
FM178223.1 - 1 - 2 85

View Full Product Details