Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC3
Entrez Gene ID
538669
Gene Name
trafficking protein particle complex 3
Gene Aliases
-
Chromosome Location
Chr.3: 109673385 - 109691951 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076120.2 NP_001069588.1 3 - 4 354 112
GenBank mRNA BC120158.1 - 3 - 4 355 112

View Full Product Details