Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GINS3
Entrez Gene ID
538943
Gene Name
GINS complex subunit 3
Gene Aliases
-
Chromosome Location
Chr.18: 26197890 - 26204585 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076136.1 NP_001069604.1 1 - 2 316 87
GenBank mRNA BC123472.1 - 1 - 2 316 87

View Full Product Details