Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIP4K2C
Entrez Gene ID
540005
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 gamma
Gene Aliases
-
Chromosome Location
Chr.5: 55886480 - 55898381 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076196.1 NP_001069664.1 3 - 4 401 90
GenBank mRNA BC120101.1 - 3 - 4 401 90

View Full Product Details