Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZCCHC12
Entrez Gene ID
540006
Gene Name
zinc finger CCHC-type containing 12
Gene Aliases
-
Chromosome Location
Chr.X: 3280803 - 3284232 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076197.1 NP_001069665.1 1 - 2 28 72
GenBank mRNA BC123686.1 - 1 - 2 28 72

View Full Product Details