Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB6
Entrez Gene ID
540218
Gene Name
zinc finger and BTB domain containing 6
Gene Aliases
ZNF482
Chromosome Location
Chr.11: 94071986 - 94096161 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076207.2 NP_001069675.1 1 - 1 1851 137
GenBank mRNA BT026250.1 - 1 - 1 1852 137

View Full Product Details