Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGDIG
Entrez Gene ID
613745
Gene Name
Rho GDP dissociation inhibitor gamma
Gene Aliases
-
Chromosome Location
Chr.25: 307142 - 309575 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076286.1 NP_001069754.1 1 - 2 124 140
GenBank mRNA BC122764.1 - 1 - 2 124 140

View Full Product Details