Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C8H9orf43
Entrez Gene ID
614006
Gene Name
chromosome 8 C9orf43 homolog
Gene Aliases
-
Chromosome Location
Chr.8: 102658618 - 102676518 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076305.2 NP_001069773.1 3 - 4 300 77
GenBank mRNA BC122744.1 - 3 - 4 301 77

View Full Product Details