Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GALNT18
Entrez Gene ID
614203
Gene Name
polypeptide N-acetylgalactosaminyltransferase 18
Gene Aliases
GALNTL4
Chromosome Location
Chr.15: 41064111 - 41419990 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076316.1 NP_001069784.1 3 - 4 784 81
GenBank mRNA BC122597.1 - 3 - 4 784 81

View Full Product Details