The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001076338.1 | NP_001069806.1 | 4 - 5 | 588 | 103 |
GenBank mRNA | BC120136.1 | - | 4 - 5 | 588 | 103 |