Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KHK
Entrez Gene ID
614868
Gene Name
ketohexokinase
Gene Aliases
-
Chromosome Location
Chr.11: 72537265 - 72550462 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076351.1 NP_001069819.1 7 - 8 848 81
GenBank mRNA BC122772.1 - 7 - 8 848 81
BT020734.1 - 6 - 7 685 81

View Full Product Details