Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
BORCS5
Entrez Gene ID
614947
Gene Name
BLOC-1 related complex subunit 5
Gene Aliases
LOH12CR1
Chromosome Location
Chr.5: 97343090 - 97440559 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076355.1 NP_001069823.1 4 - 4 713 102
GenBank mRNA BC123638.1 - 4 - 4 713 102

View Full Product Details