Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF1AD
Entrez Gene ID
615726
Gene Name
eukaryotic translation initiation factor 1A domain containing
Gene Aliases
-
Chromosome Location
Chr.29: 44080266 - 44093667 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076393.1 NP_001069861.1 3 - 4 578 69
GenBank mRNA BC120113.1 - 3 - 4 578 69
BT021815.1 - 3 - 4 535 69

View Full Product Details