Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB8
Entrez Gene ID
617269
Gene Name
ankyrin repeat and SOCS box containing 8
Gene Aliases
-
Chromosome Location
Chr.5: 32132124 - 32144541 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076452.1 NP_001069920.1 2 - 3 208 67
GenBank mRNA BC123430.1 - 2 - 3 208 67

View Full Product Details